Gene Transcripts

Gene Symbol
API5
Entrez Gene ID
8539
Gene Name
apoptosis inhibitor 5
Gene Aliases
AAC-11, AAC11
Chromosome Location
Chr. 11: 43311955 - 43344530 on Build GRCh38
UniGene ID
Hs.435771
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006718359.3 XP_006718422.1 14 3200
XM_017018464.1 XP_016873953.1 15 3257
GenBank mRNA AK021649.1 - 1 1109

View Full Product Details