Gene Transcripts
- Gene Symbol
- BTF3P9
- Entrez Gene ID
- 503543
- Gene Name
- basic transcription factor 3 pseudogene 9
- Gene Aliases
- Chromosome Location
- Chr. 1: 227433913 - 227434606 on Build GRCh38
- UniGene ID
- Hs.586511
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC062736.1 |
- |
1 |
906 |
|
View Full Product Details