Gene Transcripts

Gene Symbol
TMEM87A
Entrez Gene ID
25963
Gene Name
transmembrane protein 87A
Gene Aliases
Chromosome Location
Chr. 15: 42210452 - 42273584 on Build GRCh38
UniGene ID
Hs.511138
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110503.2 NP_001103973.1 7 1786
GenBank mRNA AK023569.1 - 1 132

View Full Product Details