Gene Transcripts
- Gene Symbol
- SHISA6
- Entrez Gene ID
- 388336
- Gene Name
- shisa family member 6
- Gene Aliases
- Chromosome Location
- Chr. 17: 11241423 - 11564063 on Build GRCh38
- UniGene ID
- Hs.592124
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL110201.1 |
- |
1 |
783 |
|
View Full Product Details