Gene Transcripts
- Gene Symbol
- PLEKHA8
- Entrez Gene ID
- 84725
- Gene Name
- pleckstrin homology domain containing A8
- Gene Aliases
- FAPP2
- Chromosome Location
- Chr. 7: 30028361 - 30130483 on Build GRCh38
- UniGene ID
- Hs.233495
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ437884.1 |
- |
1 |
152 |
|
Chr. 7 - 30081145 - 30082034 [+] |
GenBankmRNA |
CN361276.1 |
- |
1 |
135 |
|
Chr. 7 - 30081161 - 30081751 [+] |
GenBankmRNA |
CN361277.1 |
- |
|
11 |
|
Chr. 7 - 30081285 - 30082004 [+] |
View Full Product Details