Gene Transcripts
- Gene Symbol
- POU3F3
- Entrez Gene ID
- 5455
- Gene Name
- POU class 3 homeobox 3
- Gene Aliases
- BRN1, OTF8, brain-1, oct-8
- Chromosome Location
- Chr. 2: 104854068 - 104858574 on Build GRCh38
- UniGene ID
- Hs.248158
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AL534719.3 |
- |
1 |
466 |
|
Chr. 2 - 104854109 - 104855155 [+] |
GenBankmRNA |
BF929868.1 |
- |
1 |
191 |
|
Chr. 2 - 104854395 - 104854868 [+] |
GenBankmRNA |
AK125157.1 |
- |
|
460 |
|
Chr. - - [-] |
GenBankmRNA |
CN266660.1 |
- |
1 |
53 |
|
Chr. 2 - 104854524 - 104855038 [+] |
View Full Product Details