Gene Transcripts

Gene Symbol
USB1
Entrez Gene ID
79650
Gene Name
U6 snRNA biogenesis phosphodiesterase 1
Gene Aliases
C16orf57, HVSL1, Mpn1, PN, hUsb1
Chromosome Location
Chr. 16: 57999600 - 58021623 on Build GRCh38
UniGene ID
Hs.408702
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204911.1 NP_001191840.1 4 1829
GenBank mRNA AK124443.1 - 1 1623
BQ223186.1 - 1 263
BX117268.1 - 1 70

View Full Product Details