Gene Transcripts

Gene Symbol
NUP214
Entrez Gene ID
8021
Gene Name
nucleoporin 214
Gene Aliases
CAIN, CAN
Chromosome Location
Chr. 9: 131125561 - 131234670 on Build GRCh38
UniGene ID
Hs.654530
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK310515.1 - 1 30


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK128260.1 - 1 2245 Chr. 9 - 131189910 - 131194205 [+]

View Full Product Details