Gene Transcripts
- Gene Symbol
- FAM120AOS
- Entrez Gene ID
- 158293
- Gene Name
- family with sequence similarity 120A opposite strand
- Gene Aliases
- C9orf10OS
- Chromosome Location
- Chr. 9: 93446500 - 93453592 on Build GRCh38
- UniGene ID
- Hs.350364
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC015241.2 |
- |
1 |
76 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BE890203.1 |
- |
1 |
67 |
|
Chr. 9 - 93443900 - 93444654 [-] |
View Full Product Details