Gene Transcripts

Gene Symbol
FAM204A
Entrez Gene ID
63877
Gene Name
family with sequence similarity 204 member A
Gene Aliases
C10orf84, bA319I23.1
Chromosome Location
Chr. 10: 118305889 - 118342352 on Build GRCh38
UniGene ID
Hs.105575
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005270024.1 XP_005270081.1 10 4161
GenBank mRNA BX538256.1 - 1 225

View Full Product Details