Gene Transcripts
- Gene Symbol
- SFTA3
- Entrez Gene ID
- 253970
- Gene Name
- surfactant associated 3
- Gene Aliases
- NANCI, SFTPH, SP-H
- Chromosome Location
- Chr. 14: 36473288 - 36513785 on Build GRCh38
- UniGene ID
- Hs.509165
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY102071.1 |
- |
5 |
1048 |
|
View Full Product Details