Gene Transcripts
- Gene Symbol
- NFATC3
- Entrez Gene ID
- 4775
- Gene Name
- nuclear factor of activated T-cells 3
- Gene Aliases
- NFAT4, NFATX
- Chromosome Location
- Chr. 16: 68085366 - 68229259 on Build GRCh38
- UniGene ID
- Hs.436585
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK057574.1 |
- |
1 |
1431 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AI038402.1 |
- |
1 |
172 |
|
Chr. 16 - 68225971 - 68226468 [+] |
GenBankmRNA |
AI092602.1 |
- |
1 |
173 |
|
Chr. 16 - 68225971 - 68226426 [+] |
GenBankmRNA |
AI080382.1 |
- |
1 |
173 |
|
Chr. 16 - 68225971 - 68226465 [+] |
GenBankmRNA |
AA769887.1 |
- |
1 |
173 |
|
Chr. 16 - 68225971 - 68226464 [+] |
GenBankmRNA |
CA314539.1 |
- |
1 |
188 |
|
Chr. 16 - 68225968 - 68226762 [+] |
View Full Product Details