Gene Transcripts
- Gene Symbol
- AGBL4
- Entrez Gene ID
- 84871
- Gene Name
- ATP/GTP binding protein like 4
- Gene Aliases
- CCP6
- Chromosome Location
- Chr. 1: 48520157 - 50024011 on Build GRCh38
- UniGene ID
- Hs.551844
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF075049.1 |
- |
1 |
84 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
H38784.1 |
- |
1 |
84 |
|
Chr. 1 - 49690436 - 49690829 [-] |
View Full Product Details