Gene Transcripts

Gene Symbol
ZDHHC9
Entrez Gene ID
51114
Gene Name
zinc finger DHHC-type containing 9
Gene Aliases
CGI89, CXorf11, DHHC9, MMSA1, MRXSZ, ZDHHC10, ZNF379, ZNF380
Chromosome Location
Chr. X: 129803288 - 129843934 on Build GRCh38
UniGene ID
Hs.193566
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008222.2 NP_001008223.1 10 4605
NM_016032.3 NP_057116.2 11 4585


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AW969573.1 - 1 530 Chr. X - 129803308 - 129803851 [-]
GenBankmRNA CN277290.1 - 1 303 Chr. X - 129803308 - 129803588 [-]

View Full Product Details