Gene Transcripts
- Gene Symbol
- BEND5
- Entrez Gene ID
- 79656
- Gene Name
- BEN domain containing 5
- Gene Aliases
- C1orf165
- Chromosome Location
- Chr. 1: 48727523 - 48778603 on Build GRCh38
- UniGene ID
- Hs.475348
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK096573.1 |
- |
1 |
1184 |
|
View Full Product Details