Gene Transcripts
- Gene Symbol
- C2CD2
- Entrez Gene ID
- 25966
- Gene Name
- C2 calcium dependent domain containing 2
- Gene Aliases
- C21orf25, C21orf258, TMEM24L
- Chromosome Location
- Chr. 21: 41885110 - 41953957 on Build GRCh38
- UniGene ID
- Hs.473894
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK129520.1 |
- |
1 |
132 |
|
View Full Product Details