Gene Transcripts
- Gene Symbol
- BDNF-AS
- Entrez Gene ID
- 497258
- Gene Name
- BDNF antisense RNA
- Gene Aliases
- ANTI-BDNF, BDNF, BDNF-AS1, BDNFOS, NCRNA00049
- Chromosome Location
- Chr. 11: 27506852 - 27698171 on Build GRCh38
- UniGene ID
- Hs.675323
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY054391.4 |
- |
7 |
778 |
|
AY054404.4 |
- |
6 |
679 |
|
EF689023.1 |
- |
6 |
684 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK074228.1 |
- |
1 |
897 |
|
Chr. 11 - 31739916 - 31742211 [+] |
View Full Product Details