Gene Transcripts
- Gene Symbol
- Apbb2
- Entrez Gene ID
- 11787
- Gene Name
- amyloid beta (A4) precursor protein-binding, family B, member 2
- Gene Aliases
- 2310007D03Rik, FE65L1, Rirl1, TR2L, Zfra
- Chromosome Location
- Chr. 5: 66298725 - 66618817 on Build GRCm38
- UniGene ID
- Mm.5159
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK009197.1 |
- |
1 |
242 |
|
BC022590.1 |
- |
1 |
385 |
|
BC038035.1 |
- |
1 |
313 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
W82585.1 |
- |
|
153 |
|
Chr. - - [-] |
GenBankmRNA |
AA199168.1 |
- |
1 |
66 |
|
Chr. 5 - 66299005 - 66299446 [-] |
GenBankmRNA |
AA397116.1 |
- |
1 |
66 |
|
Chr. 5 - 66299068 - 66299446 [-] |
GenBankmRNA |
W75718.1 |
- |
|
49 |
|
Chr. - - [-] |
GenBankmRNA |
AA571918.1 |
- |
1 |
55 |
|
Chr. 5 - 66299116 - 66299435 [-] |
View Full Product Details