Gene Transcripts
- Gene Symbol
- Otud3
- Entrez Gene ID
- 73162
- Gene Name
- OTU domain containing 3
- Gene Aliases
- 3110030K17Rik
- Chromosome Location
- Chr. 4: 138891063 - 138913947 on Build GRCm38
- UniGene ID
- Mm.238081
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK076876.1 |
- |
1 |
153 |
|
View Full Product Details