Gene Transcripts

Gene Symbol
Tmppe
Entrez Gene ID
100504715
Gene Name
transmembrane protein with metallophosphoesterase domain
Gene Aliases
Chromosome Location
Chr. 9: 114401095 - 114411201 on Build GRCm38
UniGene ID
Mm.290516
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK082033.1 - 1 1287


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB728872.1 - 1 167 Chr. 9 - 114410771 - 114411200 [+]
GenBankmRNA BB709888.1 - 1 62 Chr. 9 - 114410876 - 114411202 [+]
GenBankmRNA BB709880.1 - 1 57 Chr. 9 - 114410881 - 114411202 [+]
GenBankmRNA CJ325705.1 - 1 200 Chr. 9 - 114410738 - 114411200 [+]
GenBankmRNA CJ325774.1 - 1 226 Chr. 9 - 114410713 - 114411200 [+]
GenBankmRNA BB726152.1 - 1 177 Chr. 9 - 114410761 - 114411202 [+]
GenBankmRNA CJ325661.1 - 1 224 Chr. 9 - 114410714 - 114411200 [+]
GenBankmRNA BY381771.1 - 1 188 Chr. 9 - 114410748 - 114411200 [+]
GenBankmRNA DV656608.1 - 1 210 Chr. 9 - 114410728 - 114411175 [+]
GenBankmRNA BE953874.1 - 1 228 Chr. 9 - 114410725 - 114411191 [+]
GenBankmRNA BB119588.1 - 1 43 Chr. 9 - 114410895 - 114411202 [+]

View Full Product Details