Gene Transcripts
- Gene Symbol
- Jakmip1
- Entrez Gene ID
- 76071
- Gene Name
- janus kinase and microtubule interacting protein 1
- Gene Aliases
- 5830437M04Rik, C330021K24Rik, Gababrbp, Marlin-1
- Chromosome Location
- Chr. 5: 37028282 - 37150614 on Build GRCm38
- UniGene ID
- Mm.85280
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK021217.1 |
- |
1 |
868 |
|
View Full Product Details