Gene Transcripts
- Gene Symbol
- Tcrg-V7
- Entrez Gene ID
- 21641
- Gene Name
- T cell receptor gamma, variable 7
- Gene Aliases
- TRGV7, Vg7
- Chromosome Location
- Chr. 13: 19178042 - 19178492 on Build GRCm38
- UniGene ID
- Mm.424521
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
Z48593.1 |
- |
1 |
231 |
|
Z48594.1 |
- |
1 |
233 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
M71214.1 |
- |
|
294 |
|
Chr. - - [-] |
View Full Product Details