Gene Transcripts
- Gene Symbol
- Qk
- Entrez Gene ID
- 19317
- Gene Name
- quaking
- Gene Aliases
- 1110003F05Rik, 1500005P18, QkI, l(17)-1Wis, l17Wis1
- Chromosome Location
- Chr. 17: 10202856 - 10319361 on Build GRCm38
- UniGene ID
- Mm.130694
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK033279.1 |
- |
1 |
1554 |
|
AK168411.1 |
- |
1 |
1709 |
|
View Full Product Details