Gene Transcripts
- Gene Symbol
- Agps
- Entrez Gene ID
- 228061
- Gene Name
- alkylglycerone phosphate synthase
- Gene Aliases
- 5832437L22, 9930035G10Rik, AW123847, Adaps, Adas, Adhaps, Adps, Aldhpsy, bs2
- Chromosome Location
- Chr. 2: 75832177 - 75931350 on Build GRCm38
- UniGene ID
- Mm.31227
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK137806.1 |
- |
1 |
2266 |
|
View Full Product Details