Gene Transcripts

Gene Symbol
Snhg1
Entrez Gene ID
83673
Gene Name
small nucleolar RNA host gene 1
Gene Aliases
Chromosome Location
Chr. 19: 8723487 - 8726326 on Build GRCm38
UniGene ID
Mm.292489
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK036616.1 - 1 1639
AK051045.1 - 2 1425


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BQ178183.1 - 2 170 Chr. 19 - 8724663 - 8726299 [+]
GenBankmRNA BQ257371.1 - 2 486 Chr. 19 - 8724356 - 8725303 [+]
GenBankmRNA AV502115.1 - 1 102 Chr. 19 - 8725029 - 8726002 [+]
GenBankmRNA CF911094.1 - 2 434 Chr. 19 - 8724403 - 8725522 [+]
GenBankmRNA CO813536.1 - 1 136 Chr. 19 - 8725004 - 8725964 [+]
GenBankmRNA AV501764.1 - 4 270 Chr. 19 - 8724162 - 8725341 [+]
GenBankmRNA U40654.1 - 1 1795 Chr. - - [-]

View Full Product Details