Gene Transcripts

Gene Symbol
Uba6
Entrez Gene ID
231380
Gene Name
ubiquitin-like modifier activating enzyme 6
Gene Aliases
4930542H01, 5730469D23Rik, AU021846, AW124799, E1-L2, Ube1l2
Chromosome Location
Chr. 5: 86110720 - 86172743 on Build GRCm38
UniGene ID
Mm.34012
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK017645.1 - 1 410
AK017687.1 - 47
AK029740.1 - 1330
AK139327.1 - 486


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CB946595.1 - 1 244 Chr. 5 - 86110013 - 86110645 [-]
GenBankmRNA CK333008.1 - 1 251 Chr. 5 - 86110246 - 86110651 [-]
GenBankmRNA BY685497.1 - 1 239 Chr. 5 - 86110246 - 86110642 [-]
GenBankmRNA CK387534.1 - 1 251 Chr. 5 - 86110246 - 86110651 [-]
GenBankmRNA AA177897.1 - 1 382 Chr. 5 - 86110348 - 86110785 [-]
GenBankmRNA BY473041.1 - 1 305 Chr. 5 - 86110250 - 86110708 [-]
GenBankmRNA CK387947.1 - 1 251 Chr. 5 - 86110090 - 86110651 [-]

View Full Product Details