Gene Transcripts

Gene Symbol
Mbd5
Entrez Gene ID
109241
Gene Name
methyl-CpG binding domain protein 5
Gene Aliases
9430004D19Rik, AA536666, AI426407, C030040A15Rik, Gm1630, OTTMUSG00000012483
Chromosome Location
Chr. 2: 48949508 - 49317070 on Build GRCm38
UniGene ID
Mm.440436
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK021136.1 - 266


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY728278.1 - 1 266 Chr. 2 - 49318853 - 49319525 [+]

View Full Product Details