Gene Transcripts
- Gene Symbol
- Clec3a
- Entrez Gene ID
- 403395
- Gene Name
- C-type lectin domain family 3, member a
- Gene Aliases
- 1110019O10Rik, Clecsf1, Gm796
- Chromosome Location
- Chr. 8: 114418034 - 114428133 on Build GRCm38
- UniGene ID
- Mm.491796
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK029455.1 |
- |
1 |
508 |
|
View Full Product Details