Gene Transcripts
- Gene Symbol
- Camkk2
- Entrez Gene ID
- 207565
- Gene Name
- calcium/calmodulin-dependent protein kinase kinase 2, beta
- Gene Aliases
- 6330570N16Rik, AW061083, mKIAA0787
- Chromosome Location
- Chr. 5: 122718580 - 122779476 on Build GRCm38
- UniGene ID
- Mm.289237
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK050846.1 |
- |
1 |
798 |
|
View Full Product Details