Gene Transcripts
- Gene Symbol
- Atpaf1
- Entrez Gene ID
- 230649
- Gene Name
- ATP synthase mitochondrial F1 complex assembly factor 1
- Gene Aliases
- 6330547J17Rik, AI593252, ATP11, ATP11p
- Chromosome Location
- Chr. 4: 115784814 - 115812314 on Build GRCm38
- UniGene ID
- Mm.109600
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK029823.1 |
- |
2 |
174 |
|
View Full Product Details