Gene Transcripts

Gene Symbol
Sfpq
Entrez Gene ID
71514
Gene Name
splicing factor proline/glutamine rich (polypyrimidine tract binding protein associated)
Gene Aliases
1110004P21Rik, 2810416M14Rik, 5730453G22Rik, 9030402K04Rik, AU021830, D4Ertd314e, Gm12940, OTTMUSG00000009329, PSF, REP1
Chromosome Location
Chr. 4: 127020560 - 127037016 on Build GRCm38
UniGene ID
Mm.257276
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK017658.1 - 1 349
AK141302.1 - 1 1643
BC033603.1 - 1 3041


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BM934843.1 - 1 478 Chr. 4 - 127034443 - 127035091 [+]
GenBankmRNA CD770415.1 - 1 312 Chr. 4 - 127034611 - 127035245 [+]
GenBankmRNA CX207240.1 - 1 435 Chr. 4 - 127034485 - 127034993 [+]

View Full Product Details