Gene Transcripts
- Gene Symbol
- Prune
- Entrez Gene ID
- 229589
- Gene Name
- prune homolog (Drosophila)
- Gene Aliases
- 9230112O05Rik, C130058A12, DRES-17, HTCD37, PRUNEM1, Prune-M1
- Chromosome Location
- Chr. 3: 95253674 - 95282076 on Build GRCm38
- UniGene ID
- Mm.14155
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020342.1 |
- |
1 |
685 |
|
AK053118.1 |
- |
|
2293 |
|
View Full Product Details