Gene Transcripts
- Gene Symbol
- FAM167A-AS1
- Entrez Gene ID
- 83656
- Gene Name
- FAM167A antisense RNA 1
- Gene Aliases
- C8orf12
- Chromosome Location
- Chr. 8: 11368402 - 11438657 on Build GRCh38
- UniGene ID
- Hs.583896
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026814.1 |
|
7 |
1034 |
|
GenBank mRNA |
AJ301563.1 |
- |
7 |
1034 |
|
BC080558.1 |
- |
7 |
1168 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000533578.4 |
- |
|
1034 |
|
Chr. 8 - 11433920 - 11438658 [+] |
GENCODE |
ENST00000624614.1 |
- |
|
1168 |
|
Chr. - - [-] |
View Full Product Details