Gene Transcripts
- Gene Symbol
- TSPEAR-AS2
- Entrez Gene ID
- 114043
- Gene Name
- TSPEAR antisense RNA 2
- Gene Aliases
- C21orf90
- Chromosome Location
- Chr. 21: 44517215 - 44518977 on Build GRCh38
- UniGene ID
- Hs.592163
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC016690.2 |
- |
2 |
1070 |
|
BC064517.1 |
- |
2 |
1070 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000354333.5 |
- |
2 |
1070 |
|
Chr. 21 - 44517617 - 44520827 [+] |
View Full Product Details