Gene Transcripts

Gene Symbol
C22orf34
Entrez Gene ID
348645
Gene Name
chromosome 22 open reading frame 34
Gene Aliases
Chromosome Location
Chr. 22: 49414524 - 49657542 on Build GRCh38
UniGene ID
Hs.133159
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017028796.1 XP_016884285.1 6 5749
GenBank mRNA AK126430.1 - 5 2781
BC048207.1 - 4 1436

Gene Symbol
ENSG00000188511
Entrez Gene ID
ENSG00000188511
Gene Name
Gene Aliases
Chromosome Location
Chr. : - on Build
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID

View Full Product Details