Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|---|
RefSeq | NR_033664.1 | 1 | 105 | |||
NR_033665.1 | 1 | 105 | ||||
GenBank mRNA | AY312367.1 | - | LIT3327 AAA1 |
1 | 105 | |
AY312368.1 | - | LIT3328 AAA1 |
1 | 105 | ||
AY312369.1 | - | LIT3332 AAA1 |
1 | 105 | ||
AY312371.1 | - | LIT3334 AAA1 |
1 | 105 | ||
AY312372.1 | - | LIT3335 AAA1 |
1 | 105 | ||
BX649082.1 | - | 1 | 68 | |||
U85653.1 | - | 1 | 68 |