Gene Transcripts

Gene Symbol
ARRDC1-AS1
Entrez Gene ID
85026
Gene Name
ARRDC1 antisense RNA 1
Gene Aliases
C9orf37
Chromosome Location
Chr. 9: 137615332 - 137618895 on Build GRCh38
UniGene ID
Hs.522520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_122035.1 2 572
NR_122036.1 2 261
GenBank mRNA AF267857.1 - 2 490
AL516492.3 - 2 572
BC011781.2 - 2 474
BC066646.1 - 2 279 5749061
BC117267.1 - 1 32 40125818
CX164735.1 - 2 261


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000496793.1 - 1 922 Chr. 9 - 137615749 - 137618906 [-]

View Full Product Details