Gene Transcripts

Gene Symbol
KU-MEL-3
Entrez Gene ID
497048
Gene Name
uncharacterized LOC497048
Gene Aliases
Chromosome Location
Chr. 6: 4610565 - 4611919 on Build GRCh38
UniGene ID
Hs.591355
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_122107.1 2 527
GenBank mRNA AY870321.1 - 2 480
BC130651.1 - 2 375 40146682
BC130655.1 - 2 380 40146686


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000380106.2 - 2 436 Chr. 6 - 4610612 - 4611918 [+]

View Full Product Details