Gene Transcripts
- Gene Symbol
- LINC00488
- Entrez Gene ID
- 677779
- Gene Name
- long intergenic non-protein coding RNA 488
- Gene Aliases
- C3orf66
- Chromosome Location
- Chr. 3: 109178165 - 109185261 on Build GRCh38
- UniGene ID
- Hs.618669
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026767.1 |
|
3 |
652 |
|
GenBank mRNA |
AK123226.1 |
- |
3 |
652 |
|
BC022563.1 |
- |
3 |
642 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG706753.1 |
- |
3 |
647 |
|
Chr. 3 - 109178173 - 109183500 [+] |
GenBankmRNA |
BI763241.1 |
- |
3 |
627 |
|
Chr. 3 - 109178190 - 109183513 [+] |
GENCODE |
ENST00000494582.1 |
- |
3 |
652 |
|
Chr. 3 - 109178165 - 109185257 [+] |
View Full Product Details