Gene Transcripts
- Gene Symbol
- PTPRD-AS2
- Entrez Gene ID
- 101929428
- Gene Name
- PTPRD antisense RNA 2 (head to head)
- Gene Aliases
- Chromosome Location
- Chr. 9: 10613206 - 10620420 on Build GRCh38
- UniGene ID
- Hs.434446
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110696.1 |
|
3 |
1219 |
|
GenBank mRNA |
BC045688.1 |
- |
3 |
1219 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000429581.2 |
- |
3 |
1223 |
|
Chr. 9 - 10613202 - 10620420 [+] |
View Full Product Details