Gene Transcripts
- Gene Symbol
- LINC01315
- Entrez Gene ID
- 102723775
- Gene Name
- long intergenic non-protein coding RNA 1315
- Gene Aliases
- lnc-C22orf32-1
- Chromosome Location
- Chr. 22: 42364400 - 42369208 on Build GRCh38
- UniGene ID
- Hs.335413
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_120595.1 |
|
2 |
537 |
|
GenBank mRNA |
BC038245.1 |
- |
2 |
503 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AI792057.1 |
- |
1 |
139 |
|
Chr. 22 - 42364498 - 42365240 [-] |
GenBankmRNA |
BI915409.1 |
- |
2 |
503 |
|
Chr. 22 - 42364694 - 42369174 [-] |
GenBankmRNA |
BF530005.1 |
- |
2 |
330 |
|
Chr. 22 - 42364640 - 42369000 [-] |
GENCODE |
ENST00000424852.1 |
- |
2 |
565 |
|
Chr. 22 - 42364525 - 42369236 [-] |
GenBankmRNA |
BX103202.1 |
- |
1 |
159 |
|
Chr. 22 - 42364650 - 42365260 [-] |
View Full Product Details