Gene Transcripts
- Gene Symbol
- ENSG00000230366
- Entrez Gene ID
- ENSG00000230366
- Gene Name
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
- Gene Symbol
- DSCR9
- Entrez Gene ID
- 257203
- Gene Name
- Down syndrome critical region 9 (non-protein coding)
- Gene Aliases
- NCRNA00038
- Chromosome Location
- Chr. 21: 37208503 - 37221736 on Build GRCh38
- UniGene ID
- Hs.505159
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ223675.1 |
- |
|
255 |
|
Chr. - - [-] |
View Full Product Details