Gene Transcripts
- Gene Symbol
- MEIS1-AS2
- Entrez Gene ID
- 100873998
- Gene Name
- MEIS1 antisense RNA 2
- Gene Aliases
- MEIS1-AS1
- Chromosome Location
- Chr. 2: 66439087 - 66440330 on Build GRCh38
- UniGene ID
- Hs.734698
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CA772438.1 |
- |
1 |
146 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000454595.1 |
- |
1 |
114 |
|
Chr. 2 - 66439443 - 66441836 [-] |
View Full Product Details