Gene Transcripts
- Gene Symbol
- C1orf64
- Entrez Gene ID
- 149563
- Gene Name
- chromosome 1 open reading frame 64
- Gene Aliases
- ERRF
- Chromosome Location
- Chr. 1: 16004236 - 16006695 on Build GRCh38
- UniGene ID
- Hs.29190
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AI732325.1 |
- |
1 |
258 |
|
AW299952.1 |
- |
1 |
269 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000437156.1 |
- |
1 |
238 |
|
Chr. 1 - 16006160 - 16006671 [-] |
View Full Product Details