Gene Transcripts
- Gene Symbol
- TMEM61
- Entrez Gene ID
- 199964
- Gene Name
- transmembrane protein 61
- Gene Aliases
- Chromosome Location
- Chr. 1: 54980614 - 54992293 on Build GRCh38
- UniGene ID
- Hs.663950
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AA918832.1 |
- |
1 |
179 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000436960.1 |
- |
1 |
164 |
|
Chr. 1 - 54980950 - 54992274 [-] |
View Full Product Details