Gene Transcripts

Gene Symbol
MAPKAPK5-AS1
Entrez Gene ID
51275
Gene Name
MAPKAPK5 antisense RNA 1
Gene Aliases
C12orf47
Chromosome Location
Chr. 12: 111839769 - 111842902 on Build GRCh38
UniGene ID
Hs.333120
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015404.1 2 2248
GenBank mRNA AK096935.1 - 2 2248
AK124158.1 - 1 2118
BC007973.2 - 3 566
BC015633.2 - 3 935
BC023547.2 - 2 1899
BC107695.1 - 2 819
JQ794500.1 - 2 2351


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000590479.5 - 4 514 Chr. 12 - 111839777 - 111841710 [-]
GENCODE ENST00000442119.5 - 3 566 Chr. 12 - 111839770 - 111841906 [-]
GENCODE ENST00000428207.4 - 2 2248 Chr. 12 - 111839769 - 111842902 [-]
GENCODE ENST00000443596.1 - 2 891 Chr. 12 - 111839767 - 111841902 [-]
GENCODE ENST00000456429.5 - 3 1919 Chr. 12 - 111839768 - 111842902 [-]

View Full Product Details