Gene Transcripts
- Gene Symbol
- ARAP1-AS2
- Entrez Gene ID
- 100506020
- Gene Name
- ARAP1 antisense RNA 2
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.503165
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125753.1 |
- |
2 |
1215 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000500163.2 |
- |
2 |
1215 |
|
Chr. 11 - 72700474 - 72705607 [+] |
View Full Product Details