Gene Transcripts
- Gene Symbol
- DLGAP1-AS2
- Entrez Gene ID
- 84777
- Gene Name
- DLGAP1 antisense RNA 2
- Gene Aliases
- Chromosome Location
- Chr. 18: 3603737 - 3610089 on Build GRCh38
- UniGene ID
- Hs.659053
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC094703.1 |
- |
2 |
1555 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000572856.1 |
- |
2 |
1555 |
|
Chr. 18 - 3603738 - 3608336 [+] |
View Full Product Details