Gene Transcripts
- Gene Symbol
- ARMCX5
- Entrez Gene ID
- 64860
- Gene Name
- armadillo repeat containing, X-linked 5
- Gene Aliases
- GASP5
- Chromosome Location
- Chr. X: 102599168 - 102604159 on Build GRCh38
- UniGene ID
- Hs.522729
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000466616.5 |
- |
5 |
701 |
|
Chr. X - 102599526 - 102650145 [+] |
GENCODE |
ENST00000476910.6 |
- |
6 |
803 |
|
Chr. X - 102599537 - 102640507 [+] |
View Full Product Details