Gene Transcripts
- Gene Symbol
- AGAP4
- Entrez Gene ID
- 119016
- Gene Name
- ArfGAP with GTPase domain, ankyrin repeat and PH domain 4
- Gene Aliases
- AGAP-4, AGAP-8, AGAP8, CTGLF1, CTGLF5, MRIP2
- Chromosome Location
- Chr. 10: 45825592 - 45858529 on Build GRCh38
- UniGene ID
- Hs.314437
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK096107.1 |
- |
1 |
16 |
|
- Gene Symbol
- BMS1P4
- Entrez Gene ID
- 729096
- Gene Name
- BMS1, ribosome biogenesis factor pseudogene 4
- Gene Aliases
- BMS1LP4
- Chromosome Location
- Chr. 10: 73699151 - 73730514 on Build GRCh38
- UniGene ID
- Hs.709171
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026592.2 |
|
12 |
1822 |
|
GenBank mRNA |
AK309545.1 |
- |
2 |
342 |
|
- Gene Symbol
- LOC107984026
- Entrez Gene ID
- 107984026
- Gene Name
- arf-GAP with GTPase, ANK repeat and PH domain-containing protein 4-like
- Gene Aliases
- Chromosome Location
- Chr. 10: 46335552 - 46838268 on Build GRCh38
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000580790.1 |
- |
12 |
1775 |
|
Chr. 10 - 73699151 - 73730487 [-] |
View Full Product Details